Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TEAD2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136953

Catalog No. NBP136953


Add to cart

Description

Description

TEAD2 Polyclonal antibody specifically detects TEAD2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

TEAD2
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000
NP_003589.1
TEA domain family member 2TEF4ETF, TEAD-2, TEF-4, transcriptional enhancer factor TEF-4
Goat
IgG
0.1 mg
Core ESC Like Genes, Stem Cell Markers
Primary
8463
Human
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
TEAD2
Peptide with sequence C-EPPDAVDSYQRH corresponding to internal region according to NP_003589.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is not expected to cross-react with other TEAD proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only