Learn More
MilliporeSigma™ anti-Tec, Polyclonal,
Manufacturer:  MilliporeSigma™ 06561
Catalog No. 06-561-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Tec
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_003215
Immunoprecipitation, Western Blot
0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
TEC; MGC126760; PSCTK4; MGC126762
Peptide containing amino acid residues 165-182 of mouse Tec
Protein A chromatography
−20°C for 24 months
Polyclonal
Human, Murine, Rat