Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tektin 3, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human TEKT3 protein.
Manufacturer: Abnova Corporation H00064518B01
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. [provided by RefSeq
Sequence: MERVGCTLTTTYAHPRPTPTNFLPAISTMASSYRDRFPHSNLTHSLSLPWRPSTYYKVASNSPSVAPYCTRSQRVSENTMLPFVSNRTTFFTRYTPDDWYRSNLTNYQESNTSRHNSEKLRVDTSRLIQDKYQQTRKTQADTTQNLGERVNDIGFWKSEIIHELDEMIGETNALTDVKKRLERALMETEAPLQVARECLFHREKRMGIDLVHDEVEAQLLTEVDTILCCQERMKLHLDKAIAQLAANRASQHELEKDLSDKQTAYRINDKCHHLRNTSDGVGYFRGVERVDATVSVPESWAKFTDDNILRSQSERAASAKLRDDIENLLVVTANEMWNQFNKVNLSFTNRIAETADAKNKIQTHLAKTLQEIFQTEMTIESIKKAIKDKTAFLKVAQTRLDERTRRPNIELCRDMAQLRLVNEVHEVDDTIQTLQQRLRDAEDTLQSLVHIKATLEYDLAVKANSLYIDQEKCMSMRKSYPNTLRLVGFCSpecifications
tektin 3 | |
Unconjugated | |
No additive | |
BC031688.1 | |
FLJ32828 | |
Mouse | |
50 uL | |
Cell Adhesion | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human TEKT3 protein. | |
TEKT3 | |
Serum | |
TEKT3 | |
TEKT3 (AAH31688.1, 1 a.a. ~ 490 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
64518 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title