Learn More
anti-Telomerase, Polyclonal, Invitrogen™
Supplier: Thermo Scientific PA138726
Description
Specifications
Telomerase | |
Rabbit | |
Affinity Purified | |
Human | |
IgG |
Polyclonal | |
Recombinant protein containing residues 174-341 of human telomerase. | |
Primary | |
4°C |