Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASD2 Rabbit anti-Human, Rat, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520439

Catalog No. PIPA520439


Add to cart

Description

Description

A suggested positive control is rat colon lysate. PA5-20439 can be used with blocking peptide PEP-0556.

Rhes, also known as tumor endothelial marker 2 (TEM2), is a small GTP-binding protein that is predomitly expressed in the striatal region of the brain. This protein belongs to the RASD subfamily of Ras-related GTP-binding protein superfamily and is thought to play a role in the normal development and function of the brain as mice lacking this gene showed increased anxiety levels and motor coordination deficits. Rhes was identified as TEM2 through analysis of genes whose expression was upregulated in tumor endothelium. Tumor endothelial markers are significantly up-regulated during angiogenesis and neoangiogenesis that are crucial for the growth of solid tumors. TEMs localized on the cell surface and conserved across species are of particular interest for future development of anti-angiogenic therapies.
Specifications

Specifications

RASD2
Polyclonal
Unconjugated
RASD2
Liquid
RASD2
A 16 amino acid peptide near the amino terminus of the human TEM2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P63033, Q96D21
MGC:4834, Rhes, TEM2; GTP-binding protein Rhes; Ras homolog enriched in striatum; tumor endothelial marker 2
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
171099, 23551
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit