Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Teprotumumab (Anti-Idiotype) Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB96341N100UG
This item is not returnable.
View return policy
Description
Teprotumumab Monoclonal specifically detects Teprotumumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
Teprotumumab | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Full-length Teprotumumab | |
100 μg | |
Primary | |
Multi-species | |
Purified |
Flow Cytometry | |
975547 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects Teprotumumab in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction