Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-TER-119, FITC, Clone: TER-119,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1435
Description
Specifically detects TER-119 Clone: TER-119 in Mouse samples, and it is validated for Flow Cytometry
The TER-119 antibody is named for the antigen to which it binds, a 52kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.Specifications
TER-119 | |
Monoclonal | |
FITC | |
Q9ES52 | |
Rat | |
Purified by size-exclusion chromatography | |
RUO | |
Primary | |
2°C to 8°C for one year from date of shipment protected from light; Do not freeze |
Flow Cytometry | |
TER-119 | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Inpp5d, 7a33, Ship, Ship1 | |
Proprietary immunogen | |
100 μg | |
Inflammation, Immunology | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction