Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TER-119, PE, Clone: TER-119,

Rat Monoclonal Antibody

$156.60 - $272.50

Specifications

Antigen TER-119
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1436MI
View Documents
MilliporeSigma™
MABF1436
25μg
Each for $156.60
Add to cart
 
MABF1437MI
View Documents
MilliporeSigma™
MABF1437
100μg
Each for $272.50
Add to cart
 
Description

Description

Specifically detects TER-119 Clone: TER-119 in Mouse samples, and it is validated for Flow Cytometry

The TER-119 antibody is named for the antigen to which it binds, a 52kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
Specifications

Specifications

TER-119
Monoclonal
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
Proprietary immunogen
Inpp5d, 7a33, Ship, Ship1
RUO
Purified by size-exclusion chromatography
Primary
Purified
TER-119
Rat
Q9ES52
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit