Learn More
TERE1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA110887
Catalog No. PIPA110887
Add to cart
Description
Description
PA1-10887 detects TERE1 in human samples. Both mouse and rat TERE1 proteins are 85% homologous to the human TERE1 protein. PA1-10887 has been successfully used in Western blot procedures, where a band of approximately 37 kDa is detected. PA1-10887 immunogen is a synthetic peptide corresponding to amino acids 32-45 of human TERE1.
A ubiquitously expressed gene designated TERE1 (Transitional Epithelial Response Gene 1), which maps to chromosome 1p36.11-1p36.33, has a significant effect on growth regulation of bladder cancer and may play an equally significant role in growth regulation of prostate cancer. The TERE1 gene is located between the micro-satellite markers D1S2667 and D1S434, a chromosome locus that has been identified as a site of a tumor suppressor gene or genes for multiple tumor types. Induced TERE1 expression in two prostate carcinoma cell lines, LNCaP and PC-3, decreased proliferation up to 80% with a significant increase in the number of cells in G1. The TERE1 transcript is present in most normal human tissues, but is reduced or absent in the majority of muscle invasive TCC (Transitional Cell Carcinoma of the bladder) tumors.
Specifications
Specifications
TERE1
Polyclonal
PBS with no preservative; pH 7.4
Q9Y5Z9
RP4-796F18.1, SCCD, TERE1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
29914
Western Blot
Unconjugated
UBIAD1
Liquid
UBIAD1
Synthetic peptide corresponding to amino acids 32-45 of human TERE1.
Protein G
RUO
Antibody
Polyclonal
Human