Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TERT, DyLight 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11089471V

Catalog No. NB11089471V

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TERT Polyclonal antibody specifically detects TERT in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TERT
1mg/mL
Western Blot, Immunocytochemistry/Immunofluorescence
O14746
EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Immunogen sequence has 84% homology to canine, rat and bovine proteins.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
TERT
Synthetic peptide made to an internal portion of human Telomerase reverse transcriptase (within residues 900-950). [Swiss-Prot# O14746]
120 kDa
0.1 ml
Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
7015
Human, Mouse, Canine
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only