Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TERT, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$361.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an internal portion of human Telomerase reverse transcriptase (within residues 900-950). [Swiss-Prot# O14746]
Antigen TERT
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11089471
View Documents
Novus Biologicals
NB11089471
0.1 ml
Each for $361.50
Add to cart
 
NB11089471T
View Documents
Novus Biologicals
NB11089471SS
0.025 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TERT Polyclonal antibody specifically detects TERT in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Rabbit
Synthetic peptide made to an internal portion of human Telomerase reverse transcriptase (within residues 900-950). [Swiss-Prot# O14746]
Polyclonal
Primary
Western Blot, Immunocytochemistry, Immunofluorescence
O14746
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
IgG
TERT
Affinity Purified
RUO
Human and mouse. Immunogen sequence has 84% homology to canine, rat and bovine proteins.
7015
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit