Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TESK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514958

Catalog No. PIPA514958

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

TESK2 is a dual specificity protein kinase activity catalyzing autophosphorylation and phosphorylation of exogenous substrates on both serine/threonine and tyrosine residues. TESK2 phosphorylates cofilin at Ser-3. It may play an important role in spermatogenesis. This nuclear enzyme is thought to be activated by autophosphorylation on Ser-219. Expression is predomitly in testis and prostate, with highest concentrations in nongerminal Sertoli cells.
Specifications

Specifications

TESK2
Polyclonal
PBS with 0.09% sodium azide
Q96S53
Testicular protein kinase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10420
Immunohistochemistry, Western Blot
Unconjugated
TESK2
Liquid
TESK2
KLH conjugated synthetic peptide between 210-240 amino acids from the central region of human TESK2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit