Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPOCK1 Monoclonal Antibody (316801), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524039
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Testican 2 or rhTestican 3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
SPOCK1 Monoclonal antibody specifically detects SPOCK1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SPOCK1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q08629 | |
SPOCK1 | |
Mouse myeloma cell line NS0-derived recombinant human Testican 1/SPOCK1 Arg22-Trp439 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
316801 | |
Unconjugated | |
SPOCK1 | |
FLJ37170; Protein SPOCK; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1; Spock; Spock1; testican; testican 1; testican-1; TIC1; TICN1 | |
Mouse | |
Protein A/G | |
RUO | |
6695 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction