Learn More
anti-Testican 1/SPOCK1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130603
Catalog No. NBP130603
Add to cart
Description
Description
Testican 1/SPOCK1 Polyclonal antibody specifically detects Testican 1/SPOCK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Testican 1/SPOCK1
LYOPH
Western Blot 1: 300-1: 2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Antisera
SPOCK1
A synthetic peptide to the c-terminal region of human Testican-1 (SPOCK) conjugated to an immunogenic carrier protein was used as the immunogen.
Unpurified
RUO
Primary
6695
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
FLJ37170, Protein SPOCK, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, SPOCKcwcv and kazal-like domains proteoglycan (testican)
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for SPOCK1.
Human, Mouse