Learn More
Testosterone Mouse anti-Chemical, Clone: 14P1F9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MIT0103
Catalog No. PIMIT0103
Add to cart
Description
Description
MIT0103 targets Testosterone in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIT0103 immunogen is testosterone-3-CMO-BSA. MIT0103 was formerly sold as a Seradyn product.
Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
Specifications
Specifications
Testosterone
Monoclonal
3.67 mg/mL
0.015M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2
Testosteron; Androderm; Testoderm; Sustanon; Mertestate; C19H28O2
Testosterone-3-CMO-BSA
Ion-exchange chromatography
RUO
Antibody
Monoclonal
ELISA, Immunoprecipitation, Radioimmune Assays
14P1F9
Unconjugated
Liquid
Mouse
IgG1
1 mg
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Chemical