Learn More
Testosterone Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175022
Catalog No. PIPA175022
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-75022 specifically detects Testosterone. PA1-75022 has been successfully used in ELISA applications. The PA1-75022 immunogen is 17b-Testosterone(3)-BSA.
Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
Specifications
Specifications
Testosterone
Polyclonal
PBS with 0.09% sodium azide
Testosteron; Androderm; Testoderm; Sustanon; Mertestate; C19H28O2
17b-Testosterone(3)-BSA
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical