Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TetR, DyLight 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178424G

Catalog No. NBP178424G


Add to cart

Description

Description

TetR Polyclonal antibody specifically detects TetR in Non-species specific samples. It is validated for Western Blot.
Specifications

Specifications

TetR
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein
A synthetic peptide made to an N-terminal portion of Escherichia coli TetR protein (between residues 1-50) [UniProt P04483]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
This antibody is used to monitor the regulation of gene expression in bacteria cells.
Western Blot
Western Blot
Affinity Purified
Rabbit
IgG
0.1 ml
Transcription Factors and Regulators
Primary
4924774
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only