Learn More
transcription factor A, mitochondrial, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TFAM protein.
Supplier: Abnova Corporation H00007019B01P
Description
This gene encodes a mitochondrial transcription factor that is a key activator of mitochondrial transcription as well as a participant in mitochondrial genome replication. Studies in mice have demonstrated that this gene product is required to regulate the mitochondrial genome copy number and is essential for embryonic development. A mouse model for Kearns-Sayre syndrome was produced when expression of this gene was eliminated by targeted disruption in heart and muscle cells. [provided by RefSeq
Sequence: MAFLRSMWGVLSALGRSGAELCTGCGSRLRSPFSFVYLPRWFSSVLASCPKKPVSSYLRFSKEQLPIFKAQNPDAKTTELIRRIAQRWRELPDSKKKIYQDAYRAEWQVYKEEISRFKEQLTPSQIMSLEKEIMDKHLKRKAMTKKKELTLLGKPKRPRSAYNVYVAERFQEAKGDSPQEKLKTVKENWKNLSDSEKELYIQHAKEDETRYHNEMKSWEEQMIEVGRKDLLRRTIKKQRKYGAEECSpecifications
transcription factor A, mitochondrial | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TFAM protein. | |
TFAM | |
MtTF1/TCF6/TCF6L1/TCF6L2/TCF6L3/mtTFA | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
7019 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003201.1 | |
TFAM | |
TFAM (NP_003192.1, 1 a.a. ∼ 246 a.a) full-length human protein. | |
50 μg | |
Cell Cycle | |
Primary | |
Human | |
Antibody |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.