Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525356

Catalog No. PIPA525356


Add to cart

Description

Description

Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization.
Specifications

Specifications

TFAP4
Polyclonal
PBS with 0.09% sodium azide
Q01664
AP-4, bHLHc41, activating enhancer-binding protein 4, class C basic helix-loop-helix protein 41, transcription factor AP-4, transcription factor AP-4 (activating enhancer-binding protein 4)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7023
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
TFAP4
Liquid
TFAP4
KLH conjugated synthetic peptide between 306-334 amino acids from the C-terminal region of human TFAP4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit