Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFB2M, Mouse, Clone: 2E10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TFB2M.
Supplier: Abnova Corporation H00064216M01
Description
Sequence: YLIQMIPRQNLFTKNLTPMNYNIFFHLLKHCFGRRSATVIDHLRSLTPLDARDILMQIGKQEDEKVVNMHPQDFKTLFETIERSKDCAYKWLYDETLEDRSpecifications
TFB2M | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022366 | |
TFB2M | |
TFB2M (NP_071761, 297 a.a. ∼ 396 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
2E10 | |
Mouse monoclonal antibody raised against a partial recombinant TFB2M. | |
TFB2M | |
FLJ22661/FLJ23182/Hkp1 | |
Mouse | |
Affinity chromatography | |
RUO | |
64216 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction