Learn More
TFE3, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127510
Description
Positive-acting transcription factor that binds to the immunoglobulin enchancer MUE3 motif. It binds also very well to a USF/MLTF site. Binding of TFE3 to DNA induces DNA binding.
PA1-27510 detects TFE3 from human, mouse, and rat samples.PA1-27510 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Transcription factor E3.Store at 4ºC short term. For extended storage aliguot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications
| TFE3 | |
| Polyclonal | |
| TFE3 | |
| RCCP2, TFEA, bHLHe33 | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 7030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| P19532, Q64092 | |
| TFE3 | |
| Synthetic peptide corresponding to human Transcription factor E3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.