Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TFG, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128762

Catalog No. NBP128762

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TFG
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
NP_001007566.1
FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene
Rabbit
IgG
0.1 ml
Signal Transduction
Primary
10342
Western Blot, Immunoprecipitation
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
TFG
The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human TRK-fused gene using the numbering given in entry NP_001007566.1 (GeneID 10342).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only