Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TFIIE beta, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132717

Catalog No. NBP132717


Add to cart

Description

Description

TFIIE beta Polyclonal antibody specifically detects TFIIE beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TFIIE beta
1.01mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
GTF2E2
Recombinant protein encompassing a sequence within the center region of human TFIIE beta. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
2961
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
FE, General transcription factor IIE subunit 2, general transcription factor IIE, polypeptide 2 (beta subunit, 34kD), general transcription factor IIE, polypeptide 2, beta 34kDa, TF2E2TFIIE-beta, TFIIE-B, transcription initiation factor IIE subunit beta
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only