Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2F1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA128259

Catalog No. PIPA128259

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28259 detects TFIIF from rat, human, mouse samples.PA1-28259 has been successfully used in Western blot applications.The PA1-28259 immunogen is: Synthetic peptide, corresponding to amino acids 1-249 of Human TFIIF.
Specifications

Specifications

TFIIF
Polyclonal
Unconjugated
GTF2F1
Purified
GTF2F1
Synthetic peptide, corresponding to amino acids 1-249 of Human TFIIF.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
1mg/mL
PBS with no preservative
P35269, Q3THK3, Q6AY96
BTF4, RAP74, TF2F1, TFIIF
Chicken
IgY
50μg
Store at -20°C
Primary
2962
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit