Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TFIP11, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140365

Catalog No. NBP140365


Add to cart

Description

Description

TFIP11 Polyclonal antibody specifically detects TFIP11 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TFIP11
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Affinity Purified
TFIP11
The immunogen for this product maps to a region between residue 100 to 150 of human tuftelin interacting protein 11 using the numbering given in entry NP_036275.1 (GeneID 24144).
Immunogen affinity purified
RUO
Primary
24144
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
bK445C9.6, DKFZP434B194, FLJ22086, hNtr1, NTR1, Septin and tuftelin-interacting protein 1, STIP, STIP-1, TIP39Spp382, tuftelin interacting protein 11, tuftelin-interacting protein 11
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only