Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526312

Catalog No. PIPA526312

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintece and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.
Specifications

Specifications

TFPT
Polyclonal
PBS with 0.09% sodium azide
P0C1Z6
FB1, INO80F, amida, INO80 complex subunit F, TCF3 fusion partner, amida, partner of the E2A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29844
Flow Cytometry, Western Blot
Unconjugated
TFPT
Liquid
TFPT
KLH conjugated synthetic peptide between 60-88 amino acids from the N-terminal region of human TFPT
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit