Learn More
TGF-beta 2 Rat anti-Mouse, Clone: 771213, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
Manufacturer:  R&D Systems MAB7346SP
 View more versions of this product
Catalog No. MAB7346SP
Add to cart
This item is not returnable. View return policy
Description
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Mouse samples. It is validated for Neutralization.
Specifications
Specifications
TGF-beta 2
771213
Neutralization 0.15-0.75 ug/mL
P27090
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
Rat
IgG2b
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TGF-beta 2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) TGF-beta 2 and rhTGF-beta 3 is observed.
Mouse
Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TGFB2
Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 2 Ala303-Ser414 Accession # P27090
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7042
Sterile PBS to a final concentration of 0.5 mg/mL.