Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 2 Rat anti-Mouse, Clone: 771213, R&D Systems™

Rat Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen TGF-beta 2
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 2 Ala303-Ser414 Accession # P27090
Clone 771213
Applications Neutralization
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB7346SP
View Documents
R&D Systems
MAB7346SP
25 ug
Each for $119.60
Add to cart
 
MAB7346500
View Documents
R&D Systems
MAB7346500
500 ug
Each for $839.00
Add to cart
 
MAB7346100
View Documents
R&D Systems
MAB7346100
100 ug
Each for $377.00
Add to cart
 
Description

Description

TGF-beta 2 Monoclonal antibody MAB7346 Specifically detects TGF-beta 2 in Mouse samples. It is validated for Neutralization
Specifications

Specifications

TGF-beta 2
771213
Unconjugated
P27090
Monoclonal
Primary
Detects mouse TGF-beta 2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) TGF-beta 2 and rhTGF-beta 3 is observed.
Protein A or G purified from hybridoma culture supernatant
Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 2 Ala303-Ser414 Accession # P27090
Neutralization
Purified
Rat
IgG2b
7042
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit