Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TGF-alpha, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145887

Catalog No. NBP145887

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

TGF-alpha
0.5 mg/mL
Western Blot, ELISA, Neutralization
NP_003227
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Rabbit
IgG
0.1 mg
Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary
7039
Western Blot, ELISA, Neutralization
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
TGFA
E. coli-expressed human TGF-α.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only