Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF alpha Sheep anti-Bovine, Human, Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA185931

Catalog No. PIPA185931


Add to cart

Description

Description

Sheep anti Human TGF alpha antibody recognizes TGF-alpha (transforming growth factor alpha) from human, rat and bovine tissues and fluids.

TGF alpha (transforming growth factor alpha) is a 50 amino acid EGF related polypeptide expressed in a range of tissues, including skin, pancreas, liver, duodenum, salivary glands, urinary and respiratory systems, and the developing and mature central nervous system (CNS). TGF-alpha is produced by monocytes, keratinocytes and neuronal cells and stimulates the proliferation of a range of epidermal and epithelial cells, through finding to the epidermal growth factor receptor (EGFR). TGF alpha is mitogenic for multipotent neural progenitors and is involved in astrogliosis.
Specifications

Specifications

TGF alpha
Polyclonal
Unconjugated
TGFA
Liquid
TGFA
Mature human recombit TGF-alpha conjugated to KLH
Ion-exchange chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Rat
ELISA
3.1 mg/mL
BBS with 0.09% sodium azide; pH 8.4
P01135, P01134
TFGA
Sheep
IgG
500 μg
4°C or -20°C if preferred
Primary
24827, 540388, 7039
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit