Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TGF-beta 2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145892

Catalog No. NBP145892


Add to cart

Description

Description

TGF-beta 2 Polyclonal antibody specifically detects TGF-beta 2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

TGF-beta 2
0.5 mg/mL
Western Blot 0.5 - 4 ug/mL
P61812
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
Rabbit
IgG
0.1 mg
Angiogenesis, Apoptosis, Cell Cycle and Replication
Primary
7042
Human, Mouse, Rat
Western Blot
Unconjugated
PBS, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Affinity Purified
TGFB2
Synthetic peptide surrounding amino acid 366 of human TGF-beta 2.
Affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
May react with pig, bovine, chick, Xenopus.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only