Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR1 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138737

Catalog No. 01-674-719


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta tissue.

A total of three members of the TGF beta family, TGFbeta1, TGFbeta2 and TGFbeta3, have been identified in mammals. Each is synthesized as a latent precursor that is subsequently cleaved forming the 112 amino acid growth factor which becomes active upon dimerization. TGFbetas mediate their activity by high affinity binding to the type II receptor 70kDa transmembrane protein with a cytoplasmic serine-threonine kinase domain.
Specifications

Specifications

TGFBR1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
P36897, P80204, Q64729
AAT5, ACVRLK4, ALK-5, ALK5, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, activin A receptor type II-like protein kinase of 53kD, activin receptor-like kinase 5, ser
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29591, 21812, 7046
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
TGFBR1
Liquid
TGFBR1
Synthetic peptide corresponding to internal region of human TGF beta R1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit