Learn More
TGFΒR Type II Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06227
Catalog No. 06-227-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect TGFβR Type II using this Anti-TGFβR Type II Antibody, NT validated for use in immunohistochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
TGFβR Type II
Unconjugated
Purified
Rabbit
IgG
400μg
Primary
NM_003242
Immunohistochemistry, Immunoprecipitation, Western Blot
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
TGFBR2; AAT3; TAAD2; TbetaR-II; TGFR-2; RIIC; MFS2; HNPCC6; FAA3; TGFbeta-RII
peptide corresponds to the first 28 N-Terminal residues of the mature human TGFb receptor
Protein G chromatography
2-8°C for 24 months
Polyclonal
Human, Murine, Rat