Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transforming growth factor beta 1 induced transcript 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human TGFB1I1 protein.

Manufacturer:  Abnova Corporation H00007041B01

Catalog No. 89-006-349


Add to cart

Description

Description

This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MPRSGAPKERPAEPLTPPPSYGHQPQTGSGESSGASGDKDHLYSTVCKPRSPKPAAPAAPPFSSSSGVLGTGLCELDRLLQELNATQFNITDEIMSQFPSSKVASGEQKEDQSEDKKRPSLPSSPSPGLPKASATSATLELDRLMASLSDFRVQNHLPASGPTQPPVVSSTNEGSPSPPEPTGKGSLDTMLGLLQSDLSRRGVPTQAKGLCGSCNKPIAGQVVTALGRAWHPEHFVCGGCSTALGGSSFFEKDGAPFCPECYFERFSPRCGFCNQPIRHKMVTALGTHWHPEHFCCVSCGEPFGDEGFHEREGRPYCRRDFLQLFAPRCQGCQGPILDNYISALSALWHPDCFVCRECFAPFSGGSFFEHEGRPLCENHFHARRGSLCATCGLPVTGRCVSALGRRFHPDHFTCTFCLRPLTKGSFQERAGKPYCQPCFLKLFG
Specifications

Specifications

transforming growth factor beta 1 induced transcript 1
Unconjugated
No additive
NM_015927.3
ARA55|HIC-5|HIC5|TSC-5
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human TGFB1I1 protein.
TGFB1I1
Serum
TGFB1I1
TGFB1I1 (NP_057011.2, 1 a.a. ~ 444 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7041
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit