Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP223668
Description
TGR5/GPBAR1 Polyclonal specifically detects TGR5/GPBAR1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.Specifications
TGR5/GPBAR1 | |
Polyclonal | |
Western Blot 1:1000 - 1:5000 | |
Q8TDU6 | |
GPBAR1 | |
A portion of amino acid 1-50 of human BG37/TGR5 was used as the immunogen. | |
0.1 mL | |
GPCR | |
151306 | |
Human, Mouse, Rat, Primate | |
Antisera |
Western Blot | |
Unconjugated | |
0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee and 72% homologous in monkey. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only