Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP11 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523050

Catalog No. PIPA523050


Add to cart

Description

Description

The target sequence has 94% sequence homology with rat and 95% sequence homology with bovine.

THAP11 is a member of the thanatos-associated proteins (THAP) family, which are zinc-dependent, sequence-specific DNA-binding factors involved in cell proliferation, apoptosis, cell cycle, chromatin modification and transcriptional regulation (Zhu, et al). THAP11 has been hypothesized to be a cell growth inhibitor via the repression of c-myc (Zhu, et al), and THAP11 expression has also been linked to the proliferation of embryonic stem cells (Dejosez M et al).
Specifications

Specifications

THAP11
Polyclonal
whole serum with 0.1% sodium azide
Q96EK4, Q9JJD0
HRIHFB2206, CTG-B43a, CTG-B45d, RONIN, THAP domain-containing protein 11
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
THAP11
Liquid
THAP11
Partial recombit human THAP11 protein expressed in E. coli.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57215, 59016
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit