Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THAP3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human THAP3 peptide using ARM Technology.
Supplier: Abnova Corporation H00090326K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
1. Customer may provide cell or tissue lysate for antibody screening.
2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
THAP3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
THAP3 | |
THAP3 | |
A synthetic peptide of human THAP3 | |
RUO | |
90326 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
THAP domain containing, apoptosis associated protein 3 | |
THAP3 | |
MGC33488 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction