Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523235

Catalog No. PIPA523235


Add to cart

Description

Description

The THEM2 gene encodes an uncharacterized hypothalamus protein and is part of the thioesterase superfamily. The thioesterase superfamily catalyzes the hydrolysis of long-chain fatty acyl-CoA thioesters. THEM2 belongs to the hotdog-fold enzyme superfamily containing a hotdog-fold and forms a back-to-back tetramer as other hotdog proteins. The protein is highly expressed in the kidney, and moderately expressed in the liver, brain, large intestine, and small intestine. Human THEM2 is co-localized with, and binds to, microtubules in all stages of the cell cycle as confirmed by immunostaining and GFP-Tag experiments. The protein is essential for eukaryotic cells growth. The gene encoding for human THEM2 is localized in the chromosomal region 6p22.2.
Specifications

Specifications

THEM2
Polyclonal
Unconjugated
ACOT13
Liquid
ACOT13
Synthetic peptide (40 CEMKVEEEHTNAIGTLHG 57) of THEM2 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9NPJ3, Q9CQR4
HT012, PNAS-27, THEM2, acyl-coenzyme A thioesterase 13, hypothalamus protein HT012, thioesterase superfamily member 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
291135, 55856, 66834
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit