Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEM2 Rabbit anti-Human, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523272

Catalog No. PIPA523272


Add to cart

Description

Description

THEM2 is a 140 amino acid enzyme with hydrolase activity belonging to the thioesterase superfamily and UPF0152 (paal) family. Human THEM2 contains a hotdog-fold and forms a back-to-back tetramer with the active site located in an inter-subunit cleft. The biological function of THEM2 is yet unknown. The role of THEM2 and Pax3 (paired box gene 3) as PC-TP-interacting proteins has been determined by using yeast two-hybrid screening. Small interference RNA silencing data suggest that the gene encoding THEM2 is essential for the cell sustained proliferation. THEM2 is highly expressed in kidney, and moderately in liver, brain, large intestine, and small intestine. THEM2 is co-localized with microtubules and is implicated in the genetic background of developmental dyslexia.
Specifications

Specifications

THEM2
Polyclonal
Unconjugated
ACOT13
Liquid
ACOT13
Synthetic peptide (123 KATGKLIAQGRHTKHLGN 140) of THEM2 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Non-human primate, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9NPJ3
HT012, PNAS-27, THEM2, acyl-coenzyme A thioesterase 13, hypothalamus protein HT012, thioesterase superfamily member 2
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
291135, 55856
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit