Learn More
Theophylline Mouse anti-Chemical, Clone: TH12-116.4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110734
Catalog No. 01-671-964
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10734 detects Theophylline. MA1-10734 has been successfully used in ELISA applications. The MA1-10734 immunogen is Theophylline, conjugated to KLH.
Theophylline is a methyl xanthine derivative from tea with diuretic, smooth muscle relaxant, bronchial dilation, cardiac and central nervous system stimulant activities. Theophylline inhibits the 3′,5′-cyclic nucleotide phosphodiesterase that degrades cyclic amp thus potentiates the actions of agents that act through adenylyl cyclases and cyclic amp. Theophylline is widely used in therapy of asthma and is not believed to cause liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=2153]
Specifications
Specifications
Theophylline
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Dimethylxanthine; 1,3-Dimethylxanthine; Elixophyllin; Theolair; Theophyllin; C7H8N4O2
Theophylline, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
TH12-116.4
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical