Learn More
anti-Thimet Oligopeptidase/THOP1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131547
Catalog No. NBP131547
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Thimet Oligopeptidase/THOP1
0.93mg/mL
Western Blot 1:1000-1:10000
Affinity Purified
THOP1
Recombinant protein encompassing a sequence within the center region of human Thimet Oligopeptidase. The exact sequence is proprietary.
79 kDa
0.1 ml
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary
7064
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rat (89%) and Bovine (89%). Not yet tested in any other species.