Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrxR1 Mouse anti-Human, Clone: 5A5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen LFMA0020

Catalog No. AML0020


Add to cart

Description

Description

A suggested positive control for this product is HeLa cells.

The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxidoreductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence (-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55-58 kilodalton in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena.
Specifications

Specifications

TrxR1
Monoclonal
Unconjugated
TXNRD1
Liquid
TXNRD1
Purified, recombit, human thioredoxin reductase 1 protein expressed in E. coli.
Ammonium sulfate precipitation
RUO
Antibody
Monoclonal
Human
ELISA, Immunoprecipitation
5A5
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide
Q16881
trxR1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7296
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit