Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

thrombopoietin, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human THPO protein.

Manufacturer:  Abnova Corporation H00007066B01

Catalog No. 89-006-355



Description

Description

Megakaryocytopoiesis is the cellular development process that leads to platelet production. The protein encoded by this gene is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene. [provided by RefSeq

Sequence: MELTELLLVVMLLLTARLTLSSPAPPACDLRVLSKLLRDSHVLHSRLSQCPEVHPLPTPVLLPAVDFSLGEWKTQMEETKAQDILGAVTLLLEGVMAARGQLGPTCLSSLLGQLSGQVRLLLGALQSLLGTQGRTTAHKDPNAIFLSFQHLLRGKVRFLMLVGGSTLCVRRAPPTTAVPSRTSLVLTLNELPNRTSGLLETNFTASARTTGSGLLKWQQGFRAKIPGLLNQTSRSLDQIPGYLNRIHELLNGTRGLFPGPSRRTLGAPDISSGTSDTGSLPPNLQPGYSPSPTHPPTGQYTLFPLPPTLPTPVVQLHPLLPDPSAPTPTPTSPLLNTSYTHSQNLSQEG
Specifications

Specifications

thrombopoietin
Unconjugated
No additive
NM_199228.1
MGC163194|MGDF|MKCSF|ML|MPLLG|TPO
Mouse
50 uL
Cytokines
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human THPO protein.
THPO
Serum
THPO
THPO (AAI30323.1, 1 a.a. ~ 349 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7066
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit