Learn More
anti-Thrombomodulin/BDCA-3, APC, Clone: 461714, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB3894A
Catalog No. FAB3894A
Add to cart
This item is not returnable. View return policy
Description
Description
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Thrombomodulin/BDCA-3
461714
Flow Cytometry 10 uL/10^6 cells
P15306
BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM
Rat
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse Thrombomodulin/BDCA-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Thrombomodulin is observed.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
THBD
Mouse myeloma cell line NS0-derived recombinant mouse Thrombomodulin/BDCA-3 Leu17-Ser517 Accession # P15306
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7056
Mouse