Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-1 Mouse anti-Human, Clone: 301221, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen Thrombospondin-1
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-1 Asn19-Pro1170 Accession # CAA32889
Clone 301221
Applications Western Blot, Immunoprecipitation, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB3074SP
View Documents
R&D Systems
MAB3074SP
25 ug
Each for $114.50
Add to cart
 
MAB3074100
View Documents
R&D Systems
MAB3074100
100 ug
Each for $377.00
Add to cart
 
MAB3074500
View Documents
R&D Systems
MAB3074500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Thrombospondin-1 Monoclonal antibody MAB3074 Specifically detects Thrombospondin-1 in Human samples. It is validated for ELISA Capture, Immunoprecipitation, Western Blotting
Specifications

Specifications

Thrombospondin-1
301221
Unconjugated
CAA32889
Monoclonal
Primary
Detects human Thrombospondin-1 in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant human (rh) Thrombospindin-2 or rhThrombospondin-4 is observed.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-1 Asn19-Pro1170 Accession # CAA32889
Western Blot, Immunoprecipitation, ELISA
Purified
Mouse
IgG2b
7057
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit