Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Thrombospondin-2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 244200020.1MG

Catalog No. 24-420-00201

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Thrombospondin-2
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
THBS2
This antibody is specific for the N Terminus Region of the target protein.
Immunogen affinity purified
RUO
Primary
7058
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P35442
thrombospondin 2, TSP2thrombospondin-2
Rabbit
IgG
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human THBS2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only