Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Thrombospondin-3, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136996

Catalog No. NBP136996


Add to cart

Description

Description

Thrombospondin-3 Polyclonal antibody specifically detects Thrombospondin-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

Thrombospondin-3
0.5 mg/mL
Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Peptide ELISA Detection limit 1:32000
NP_009043.1
MGC119565, thrombospondin 3, thrombospondin-3, TSP3MGC119564
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific to the C Terminus.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
THBS3
Peptide with sequence CNDTVPEDFEPFRRQ corresponding to C-Terminus according to NP_009043.1.
Immunogen affinity purified
RUO
Primary
7059
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only