Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM59 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521575

Catalog No. PIPA521575


Add to cart

Description

Description

PA5-21575 targets thymic dendritic cell-derived factor 1 precursor in WB and IHC(P) applications and shows reactivity with Human, Rat and mouse samples. The PA5-21575 immunogen is a recombit fragment corresponding to a region within amino acids 16 and 311 of TMEM59 (Uniprot IDNo.Q9BXS4). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

TMEM59
Polyclonal
Unconjugated
TMEM59
Liquid
TMEM59
Recombit fragment corresponding to a region within amino acids 16 and 311 of TMEM59 (Uniprot IDNo.Q9BXS4)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9BXS4, Q9QY73
RP4-758J24.2, C1orf8, HSPC001, liver membrane-bound protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100196907, 56374, 9528
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit