Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thyroglobulin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8306
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Thyroglobulin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AITD3, AITD3TGN, cog, TDH3, Tg, TGN, thyroglobulin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7038 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry | |
904802 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P01266 | |
TG | |
Chinese hamster ovary cell line CHO-derived recombinant human Thyroglobulin Met1-Lys2768 Accession # P01266 | |
100 μg | |
Primary | |
Detects human Thyroglobulin in direct ELISAs. | |
Human | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction