Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Mouse anti-Human, Clone: SPM517, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA138758

Catalog No. 01672654

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

A suggested positive control is thyroid tissue.

Antibody to thyroglobulin has been shown to be useful in positive ifentification of thyroid carcinomas of the papillary and follicular types. Demonstration of thytoglobulin in a metastatic lesion establishes the thyroid origin of the tumor. Adenocarcinomas of non-thyroid origin are not reactive.
Specifications

Specifications

Thyroglobulin
Monoclonal
Unconjugated
TG
Liquid
TG
Human thyroglobulin
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7038
Immunohistochemistry (Paraffin)
SPM517
tissue culture supernatant diluted in TBS with 1% BSA and 0.099% sodium azide
P01266
AITD3, TGN
Mouse
IgG1
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit